Skip to Content
Merck
All Photos(1)

Key Documents

34375

Supelco

Thiodicarb

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N4O4S3
CAS Number:
Molecular Weight:
354.47
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CS\C(C)=N/OC(=O)N(C)SN(C)C(=O)O\N=C(\C)SC

InChI

1S/C10H18N4O4S3/c1-7(19-5)11-17-9(15)13(3)21-14(4)10(16)18-12-8(2)20-6/h1-6H3/b11-7-,12-8-

InChI key

XDOTVMNBCQVZKG-OXAWKVHCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiodicarb is a carbamate herbicide effective against Lepidoptera, Coleoptera, Diptera and Hemiptera pests. It acts by inhibiting acetylcholinesterase activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiodicarb may be used as an analytical reference standard for the determination of the analyte in cotton, fruits/vegetables, and in different food matrices by various chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of carbamate pesticide residues in vegetables and fruits by liquid chromatography- atmospheric pressure photoionization- mass spectrometry and atmospheric pressure chemical ionization- mass spectrometry.
Takino M, et al.
Journal of Agricultural and Food Chemistry, 52(4), 727-735 (2004)
Simultaneous determination of thiodicarb and its main metabolite residues in cotton by ultra-performance liquid chromatography coupled to tandem mass spectrometry.
Wu Y, et al.
Analytical Methods : Advancing Methods and Applications, 5(4), 1052-1057 (2013)
D S Brickle et al.
Journal of economic entomology, 94(1), 86-92 (2001-03-10)
Six insecticides of different chemistries were evaluated against the cotton bollworm, Helicoverpa zea (Boddie), in non-B.t. (Deltapine 'DP 5415', Deltapine 'DP 5415RR') and transgenic Bacillus thuringiensis (Berliner) (B.t.) (Deltapine 'NuCOTN 33B', Deltapine 'DP 458 B/RR') cotton. In 1998, treatments consisted
Sally K Moccelini et al.
Talanta, 82(1), 164-170 (2010-08-06)
A biosensor based on alfalfa sprout (Medicago sativa) homogenate as a source of peroxidase is proposed for the determination of thiodicarb by square-wave voltammetry. This enzyme was immobilized in self-assembled monolayers of l-cysteine on a gold electrode. Several parameters were
C L Lanning et al.
Toxicology letters, 85(3), 127-133 (1996-06-01)
Pesticides have been shown to interact with the multidrug resistance protein associated with cancer chemotherapy, P-glycoprotein (P-gp). P-gp, therefore, has also been implicated in the development of pesticide resistance. The purpose of this study was to characterize the effect P-gp

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service