Skip to Content
Merck
All Photos(1)

Key Documents

33823

Supelco

Decoquinate

VETRANAL®, analytical standard

Synonym(s):

Ethyl 6-decyloxy-7-ethoxy-4-hydroxy-3-quinolinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H35NO5
CAS Number:
Molecular Weight:
417.54
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCOc1cc2c(O)c(cnc2cc1OCC)C(=O)OCC

InChI

1S/C24H35NO5/c1-4-7-8-9-10-11-12-13-14-30-21-15-18-20(16-22(21)28-5-2)25-17-19(23(18)26)24(27)29-6-3/h15-17H,4-14H2,1-3H3,(H,25,26)

InChI key

JHAYEQICABJSTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decoquinate is a quinolone coccidiostat used in poultry, to retard the growth of coccidian parasites and can also show activity against Plasmodium falciparum parasite, which causes malaria in humans.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytochrome b mutation identified in a decoquinate-resistant mutant of Toxoplasma gondii.
D C McFadden et al.
The Journal of eukaryotic microbiology, 46(5), 81S-82S (1999-10-16)
A chemical genomic analysis of decoquinate, a Plasmodium falciparum cytochrome b inhibitor
Nam T-G, et al.
ACS Chemical Biology, 6, 1214-1222 (2011)
M A Taylor et al.
Journal of veterinary pharmacology and therapeutics, 35(5), 417-427 (2012-06-26)
Decoquinate is a quinolone derivative that has been used for over 20 years in the control of coccidiosis in domestic ruminants. Decoquinate treats coccidiosis in lambs and calves and prevents coccidiosis in lambs when administered in feed at a dosage
M Lallemand et al.
The Veterinary record, 159(20), 672-676 (2006-11-14)
Ninety, seven- to 10-day-old calves were allocated to three groups of 30 and treated daily for seven days with either 100 microg/kg halofuginone hydrobromide or 2.5 mg/kg decoquinate orally or left untreated as controls. The levels of diarrhoea and dehydration
Medicating feeds for early lambing ewes.
Neville Suttle
The Veterinary record, 170(6), 161-161 (2012-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service