Skip to Content
Merck
All Photos(1)

Key Documents

31618

Supelco

2-Naphthylamine

analytical standard

Synonym(s):

β-Naphthylamine, 2-Aminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NH2
CAS Number:
Molecular Weight:
143.19
Beilstein:
3939429
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

306 °C (lit.)

mp

111-113 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Nc1ccc2ccccc2c1

InChI

1S/C10H9N/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,11H2

InChI key

JBIJLHTVPXGSAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Naphthylamine is a carcinogenic aromatic amine which induces tumor in urinary bladder.

Application

2-Naphthylamine has been used as reference standard during isolation and identification of urinary metabolites using preparative thin-layer chromatography and gas-liquid chromatography/mass spectral analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Formation of urothelial and hepatic DNA adducts from carcinogen 2-naphthylamine.
Kadlubar FF
Carcinogenesis, 2(5), 467-470 (1981)
Metabolic conversion of 1-and 2-nitronaphthalene to 1-and 2-naphthylamine in the rat.
Johnson, Dale E., and Herbert H. Cornish.
Toxicology and Applied Pharmacology, 46.3, 549-553 (1978)
Carlos de la Haba et al.
Biochimica et biophysica acta, 1838(1 Pt B), 148-157 (2013-08-21)
Receptor-ligand binding is an essential interaction for biological function. Oxidative stress can modify receptors and/or membrane lipid dynamics, thus altering cell physiological functions. The aim of this study is to analyze how oxidative stress may alter receptor-ligand binding and lipid
Carlos de la Haba et al.
Biochimica et biophysica acta, 1828(2), 357-364 (2012-09-04)
Plasma membrane is one of the preferential targets of reactive oxygen species which cause lipid peroxidation. This process modifies membrane properties such as membrane fluidity, a very important physical feature known to modulate membrane protein localization and function. The aim
Dylan M Owen et al.
Nature communications, 3, 1256-1256 (2012-12-06)
Lipid microdomains are postulated to regulate many membrane-associated processes but have remained highly controversial. Here we provide the first direct evidence that the plasma membrane of intact, live cells is comprised of a sub-resolution mixture of approximately 76% ordered and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service