Skip to Content
Merck
All Photos(4)

Key Documents

254002

Sigma-Aldrich

Pyrogallol

ACS reagent

Synonym(s):

1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
Beilstein:
907431
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

4.4 (vs air)

vapor pressure

10 mmHg ( 167.7 °C)

form

solid

ign. residue

≤0.005%

bp

309 °C (lit.)

mp

133-134 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤5 ppm

SMILES string

Oc1cccc(O)c1O

InChI

1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H

InChI key

WQGWDDDVZFFDIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Metal-complexing agent
Pyrogallol undergoes quick autooxidation in alkaline solution. Due to this property, its alkaline solution is generally used for the removal of oxygen from gases. Pyrogallol may also be used to prepare pyrogallol monomethyl ether via methylation and gallacetophenone via a modified Nencki reaction.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrogallol 1-monomethyl ether
Surrey AR
Organic Syntheses, 26, 90-90 (1946)
Involvement of the superoxide anion radical in the autoxidation of pyrogallol and a convenient assay for superoxide dismutase.
Marklund S and Marklund G.
European Journal of Biochemistry, 47(3), 469-474 (1974)
Sanja Matić et al.
Mutation research, 755(2), 81-89 (2013-07-09)
The present study was undertaken to investigate the hepatoprotective effect of the methanol extract of Cotinus coggygria Scop. in rats exposed to the hepatotoxic compound pyrogallol. Assessed with the alkaline version of the comet assay, 1000 and 2000mg/kg body weight
Gianluca Dimartino
Journal of AOAC International, 90(5), 1340-1345 (2007-10-25)
A convenient method is presented for determination of vitamin D in natural cheese, processed cheese, milk, cereals, noncarbonated soft drinks, and juice by liquid chromatography/ mass spectrometry (LC/MS). Samples were saponified, extracted, evaporated, redissolved in acetonitrile, and injected into an
Gallacetophenone
Badhwar IC and Venkataraman K
Organic Syntheses, 14, 40-40 (1934)

Protocols

To standardize a procedure for the assay of Peroxidase using 2,2'-Azino-bis(3-Ethylbenzthiazoline-6-Sulfonic Acid) as a substrate.

This procedure is for the determination of Peroxidase enzymatic activity using Pyrogallol as the substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service