Skip to Content
Merck
All Photos(2)

Key Documents

MABT826

Sigma-Aldrich

Anti-Actin Antibody, clone 2G2

clone 2G2, from mouse

Synonym(s):

Actin, alpha skeletal muscle, Actin, Alpha-actin-1, G-actin, Nuclear actin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
2G2, monoclonal
application:
ICC
WB
species reactivity:
rat, human, frog, rabbit, mouse, chicken, monkey
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
3

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G2, monoclonal

species reactivity

rat, human, frog, rabbit, mouse, chicken, monkey

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actin, alpha skeletal muscle (UniProt P68135; also known as Alpha-actin-1) is encoded by the ACTA1 (also known as ACTA) gene (Gene ID 100009506) in Oryctolagus cuniculus (Rabbit) species. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt. Research indicates distinct actin conformations/structures in the nucleus and the cytoplasm of different cell types and that their distribution varies in response to external signals. In additon to serving as the basic building blocks of cytoskeletal microfilaments, actins are involved in multiple nuclear functions and adopt specific conformations that are important for their association with nuclear actin-binding proteins.

Specificity

Clone 2G2 recognizes a tripartite epitope identified in a region of the actin monomer (G-actin) that is not exposed on the surface of filamentous F-actin, but is accessible on the surface of profilin-complexed actin molecule (Gonsior, S.M., et al. (1999). J. Cell Sci. 112 (Pt 6):797-809).

Immunogen

Epitope: Nonsequential epitope formed by regions within a.a. 104-226.
Reconstituted complex of rabbit skeletal muscle actin and calf thymus profilin.

Application

Anti-Actin Antibody, clone 2G2 is an antibody against Actin for use in Immunocytochemistry, Western Blotting.
Immunocytochemistry Analysis: 5.0 µg/mL from a representative lot detected Actin in serum-starved C2C12 mouse myoblasts.
Immunocytochemistry Analysis: A representative lot showed fixation-dependent cellular staining by fluorescent immunocytochemistry. Clone 2G2 stained nuclear dot-like structures, but not cytoplasmic myofibrils and stress-fibers (MF & SF) in formaldehyde-fixed chicken and mouse myogenic cells, while MF & SF, but not nuclei staining was seen in methanol-fixed chicken cardiac myocytes, fibroblasts, and in vitro differentiated chicken skeletal muscle myotubes (Gonsior, S.M., et al. (1999). J. Cell Sci. 112 (Pt 6):797-809).
Immunocytochemistry Analysis: A representative lot detected nuclear actin immunoreactivity in formaldehyde-fixed COS and NIH-3T3 fibrobastic cells, normal rat kidney NRK epithelial cells, and Xenopus oocytes by fluorescent immunocytochemistry (Gonsior, S.M., et al. (1999). J. Cell Sci. 112 (Pt 6):797-809).
Immunocytochemistry Analysis: A representative lot, when microjected into FH12 rat fibroblasts prior to formaldehyde fixation, showed dot-like staining of cytoplasmic actin monomer (G-actin), but not the microfilament (e.g. myofibrils and stress-fibers) structures (F-actin) seen with FITC-phalloidin staining (Gonsior, S.M., et al. (1999). J. Cell Sci. 112 (Pt 6):797-809).
Immunocytochemistry Analysis: A representative lot detected subnuclear compartmentalized ss-actin immunoreactive foci in onion (Allium cepa) cells by immunofluorescent confocal microscopy (Cruz, J.R., et al. (2009). Chromosoma. 118(2):193-207).
Immunocytochemistry Analysis: A representative lot detected actin immunoreactivity in the nucleus, in the submembraneous actin web, and at the filopodia tips by fluorescent immunocytochemistry staining of fixed rat fibroblasts and HeLa cells (Jockusch, B.M., et al. (2006). Trends Cell Biol. 16(8):391-396).
Western Blotting Analysis: A representative lot showed a broad-spectrum species (rabbit, chicken, mouse) reactivity against SDS-denatured actins (alpha, beta and gamma) by Western blotting. However, clone 2G2 displayed no reactivity toward slime mold (Physarum polycephalum) actin or actins from muscle tissues of several invertebrate species (Gonsior, S.M., et al. (1999). J. Cell Sci. 112 (Pt 6):797-809).
Western Blotting Analysis: A representative lot detected actin in all soluble and insoluble fractions of onion (Allium cepa) nuclei preparations (Cruz, J.R., et al. (2009). Chromosoma. 118(2):193-207).
Western Blotting Analysis: A representative lot detected an internal actin fragment corresponding to a.a. 104-226 of rabbit skeletal muscle alpha-actin obtained by limited proteolytic digestion (Gonsior, S.M., et al. (1999). J. Cell Sci. 112 (Pt 6):797-809).
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Immunocytochemistry in serum-starved HeLa cells.

Immunocytochemistry Analysis: 5.0 µg/mL of this antibody detected Actin in serum-starved HeLa cells.

Target description

~42 kDa calculated

Physical form

Format: Purified
Purified mouse monoclonal IgMκ antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulina Ormazabal et al.
International journal of obesity (2005), 42(12), 2012-2021 (2018-05-18)
The occurrence of chronic inflammation in visceral adipose tissue (VAT) in obese subjects precipitates the development of insulin resistance and type 2 diabetes (T2D). Anthocyanins and their main metabolite protocatechuic acid (PCA) have been demonstrated to stimulate insulin signaling in
Katja Burk et al.
Scientific reports, 7(1), 2149-2149 (2017-05-21)
The sorting of activated receptors into distinct endosomal compartments is essential to activate specific signaling cascades and cellular events including growth and survival. However, the proteins involved in this sorting are not well understood. We discovered a novel role of
Nathalie Journiac et al.
Cell reports, 31(2), 107506-107506 (2020-04-16)
A distinctive feature of neocortical development is the highly coordinated production of different progenitor cell subtypes, which are critical for ensuring adequate neurogenic outcome and the development of normal neocortical size. To further understand the mechanisms that underlie neocortical growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service