Skip to Content
Merck
All Photos(1)

Key Documents

MABT207

Sigma-Aldrich

Anti-Integrin αV (ITGAV/CD51) Antibody

mouse monoclonal, 272-17E6

Synonym(s):

Integrin alpha-V, Vitronectin receptor subunit alpha, CD51, Integrin alpha-V heavy chain, Integrin alpha-V light chain, Intergrin av

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin alpha-V Antibody, clone 272-17E6 (Azide free), clone 272-17E6, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

272-17E6, monoclonal

species reactivity

human

technique(s)

activity assay: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

Integrin alpha-V (or CD51; vitronectin receptor subunit alpha) protein is a transmembrane receptor that mediates adhesion of cells to each other and to the extracellular matrix. Integrin receptors are activated by a variety of intracellular signals that modify its cytoplasmic domain and induce configural changes in integrin molecules thereby enabling ligand binding to the extracellular surface. An important part of this process is the binding of the actin protein, talin, to the integrin cytoplasmic domain.

Immunogen

Human Placental ɑvβ3

Application

Anti-Integrin alpha-V Antibody, clone 272-17E6 (Azide free) detects level of Integrin alpha-V & has been published & validated for use in Flow Cytometry, Immunoprecipitation, Activity Assay.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to detect Integrin alpha-V in M21 cell lysates. (Mitjans, F., et al (1995). Journal of Cell Science. 108:2825-2838.)
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Flow Cytometry in HeLa cells.

Flow Cytometry Analysis: 0.5 µg of this antibody detected Integrin alpha-V in HeLa cells.

Target description

116 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservative.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khalil Zaman et al.
International journal of cancer, 118(3), 755-764 (2005-08-23)
Anti-angiogenic therapies are currently in cancer clinical trials, but to date there are no established tests for evaluating the angiogenic status of a patient. We measured 11 circulating angiogenesis-associated molecules in cancer patients before and after local treatment. The purpose
Daniel V Bax et al.
The Journal of biological chemistry, 284(42), 28616-28623 (2009-07-21)
Elastin fibers are predominantly composed of the secreted monomer tropoelastin. This protein assembly confers elasticity to all vertebrate elastic tissues including arteries, lung, skin, vocal folds, and elastic cartilage. In this study we examined the mechanism of cell interactions with
Pengcheng Li et al.
Cell adhesion & migration, 14(1), 1-11 (2020-01-08)
Reducted arylamine N-acetyltransferase (NAT1) in breast cancers is associated with poor patient survival. NAT1 has also been associated with changes in cancer cell survival and invasion both invitro and invivo. Here, we report the effects of NAT1 in cancer cell
Maria Rita Milone et al.
Oncotarget, 6(7), 5324-5341 (2014-12-08)
Proteomic analysis identified differentially expressed proteins between zoledronic acid-resistant and aggressive DU145R80 prostate cancer (PCa) cells and their parental DU145 cells. Ingenuity Pathway Analysis (IPA) showed a strong relationship between the identified proteins within a network associated with cancer and
Miao Yin et al.
Oncotarget, 8(58), 98280-98297 (2017-12-13)
Malignant mesothelioma originates from mesothelial cells and is a cancer type that aggressively invades into the surrounding tissue, has poor prognosis and no effective treatment. Gremlin-1 is a cysteine knot protein that functions by inhibiting BMP-pathway activity during development. BMP-independent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service