Skip to Content
Merck
All Photos(1)

Key Documents

MAB13444

Sigma-Aldrich

Anti-Integrin α6 Antibody, clone BnC6B4

clone BnC6B4, Chemicon®, from mouse

Synonym(s):

CD49f

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
BnC6B4, monoclonal
application:
ICC
IHC
IP
WB
species reactivity:
chicken
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BnC6B4, monoclonal

species reactivity

chicken

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

Specificity

Reactive with chicken integrin alpha6, as demonstrated by experiments with cDNA-transfected NIH3T3 cells followed by immunoblots.

Immunogen

Integrin alpha subunits purified from adult chicken brain tissue, denatured in 6M guanidine isothiocyanate, and depleted of beta1 subunit.

Application

Anti-Integrin α6 Antibody, clone BnC6B4 is an antibody against Integrin α6 for use in IP, WB, IC, IH.
Western blot

Immunoprecipitation: Precipitates the monomeric alpha6 subunit after the dimeric receptor has been denatured, but does not precipitate the native receptor.

Immunohistochemistry: Effective for immunohistochemistry of methanol-fixed cryosections. An overnight methanol fix is suggested. Previous experiments have used 14 micron sections of chicken embryonic tissue. Typical antibodies dilutions are 35 μg/mL, with overnight antibody incubation, followed by detection with a goat anti-mouse Ig -FITC secondary antiobody.

Immunocytochemistry: Effective for methanol-fixed cultured cells (15 minute fixation).

Immunoblot: Recognizes a 130 kDa band in chicken brain tissue and a 140 kDa band in transfected NIH3T3 cells (the difference in molecular weight is likely due to differential glycosylation).

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren R Strachan et al.
The Journal of cell biology, 167(3), 545-554 (2004-11-03)
Cell migration is essential for proper development of numerous structures derived from embryonic neural crest cells (NCCs). Although the migratory pathways of NCCs have been determined, the molecular mechanisms regulating NCC motility remain unclear. NCC migration is integrin dependent, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service