HLINJMAG-75K
MILLIPLEX® Human Liver Injury Magnetic Bead Panel - Toxicity Multiplex Assay
About This Item
Recommended Products
description
The analytes available for this multiplex kit are: Liver-type arginase 1 (ARG1), α-glutathianone S-transferase (GSTα), malate dehydrogenase 1 (MDH1), sorbitol dehydrogenase (SDH), and 5′-Nucleotidase (5′-NT/CD73).
Quality Level
100
200
species reactivity
human
manufacturer/tradename
Milliplex®
assay range
accuracy: 106-123%
standard curve range: 14-10,000 pg/mL
(GSTα)
standard curve range: 41-30,000 pg/mL
(5′-NT/CD73)
standard curve range: 549-400,000 pg/mL
(SDH)
standard curve range: 69-50,000 pg/mL
(ARG1)
standard curve range: 69-50,000 pg/mL
(MDH1)
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex xMAP)
shipped in
ambient
General description
The MILLIPLEX® Human Liver Injury Panel contains all the components necessary to simultaneous quantify the following 5 analytes in serum and plasma samples:
• Liver-Type Arginase 1 (ARG1)*
• Malate dehydrogenase 1 (MDH1)*
• α-glutathione S-transferase (GSTα)*
• Sorbitol Dehydrogenase (SDH)*
• 5′-Nucleotidase/CD73 (5′-NT)
*ARG1, GSTα, and SDH are biomarkers listed in the Predictive Safety Testing Consortium (PSTC) project pipeline which have a strong translational role in drug safety testing.
The MILLIPLEX® portfolio offers the broadest selection of analytes across a wide range of disease states and species. Once the analytes of interest have been identified, you can rely on the quality that we build into each kit to produce results you can trust. In addition to the assay characteristics listed in the protocol, other performance criteria evaluated during the validation process include: cross-reactivity, dilution linearity, kit stability, and sample behavior (e.g. detectability and stability).
Panel Type: Toxicity
Specificity
There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes within a panel.
Application
- Analytes: ARG1, GSTα, SDH 5′-NT/CD73, Malate dehydrogenase 1 (MDH1)
- Recommended Sample type: serum, plasma, and tissue culture supernatants
- Recommended Sample dilution: 1:5 serum or plasma samples. Tissue culture supernatants may require dilution as well.
- Assay Run Time: One day or Overnight
- Research Category: Toxicity
Features and Benefits
Packaging
Storage and Stability
Other Notes
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.
MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.
MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.
MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service