Skip to Content
Merck
All Photos(1)

Key Documents

AP187P

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed

0.8 mg/mL, Chemicon®

Synonym(s):

Goat Anti-Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

0.8 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Rabbit IgG (H+L) (absorbed for minimal cross-reactivity to Human, Mouse and Rat Serum Proteins)

Application

Goat anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed is an antibody against Rabbit IgG for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
Suggested dilution for most applications:

1:500-1:5,000 for Immunoohistochemistry on tissue sections.

1:5,000-1:100,000 for ELISA and Western blotting with chromogenic substrates.

1:10,000-1:200,000 for western blotting with chemiluminescent substrates.

Optimal working dilutions must be determined by the end user.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA.

RECONSTITUTION:

Reconstitute with 1.5 mL sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander A Dolskiy et al.
BioMed research international, 2017, 3582601-3582601 (2017-12-07)
Fragile X syndrome is the most common cause of inherited intellectual disability in humans. It is a result of CGG repeat expansion in the 5' untranslated region (5' UTR) of the FMR1 gene. This gene encodes the FMRP protein that
Shuguo Yang et al.
Molecular medicine reports, 18(6), 5207-5214 (2018-10-03)
It has been well established that long non‑coding RNAs (lncRNAs) are crucial mediators in a diverse range of diseases, including atherosclerosis. The present study aimed to examine the molecular mechanisms underlying the association between steroid receptor RNA activator (SRA) and
Lei Guo et al.
Molecular medicine reports, 15(4), 2241-2246 (2017-03-06)
The aim of the present study was to determine whether amyloid-β (Aβ) protein could be detected in the serum and cerebrospinal fluid (CSF) of rats with traumatic spinal cord injury (SCI) and whether Aβ protein levels in serum and CSF
Joyce J W Wong et al.
Nature communications, 8(1), 781-781 (2017-10-05)
Nipah virus is an emergent paramyxovirus that causes deadly encephalitis and respiratory infections in humans. Two glycoproteins coordinate the infection of host cells, an attachment protein (G), which binds to cell surface receptors, and a fusion (F) protein, which carries
Jun Gui et al.
Nature cancer, 1(6), 603-619 (2021-06-15)
Primary tumor-derived factors (TDFs) act upon normal cells to generate a pre-metastatic niche, which promotes colonization of target organs by disseminated malignant cells. Here we report that TDFs-induced activation of the p38α kinase in lung fibroblasts plays a critical role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service