Skip to Content
Merck
All Photos(1)

Key Documents

8.52090

Sigma-Aldrich

Fmoc-Tyr(PO(NMe₂)₂)-OH

Novabiochem®

Synonym(s):

Fmoc-Tyr(PO(NMe₂)₂)-OH, N-α-Fmoc-O-(bis-dimethylamino-phosphono)-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N3O6P
CAS Number:
Molecular Weight:
537.54
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥93.0% (acidimetric)
≥96% (TLC)
≥96.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

[P](=O)(N(C)C)(N(C)C)Oc1ccc(cc1)CC(NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O

InChI

1S/C28H32N3O6P/c1-30(2)38(35,31(3)4)37-20-15-13-19(14-16-20)17-26(27(32)33)29-28(34)36-18-25-23-11-7-5-9-21(23)22-10-6-8-12-24(22)25/h5-16,25-26H,17-18H2,1-4H3,(H,29,34)(H,32,33)

InChI key

WBUXJDYGAMFMPS-UHFFFAOYSA-N

General description

Fmoc-Tyr(PO(NMe2)2)-OH is a useful derivative for the synthesis of phosphotyrosine-containing peptides by Fmoc SPPS [1]. As the side-chain phosphate group is fully protected, this derivative is compatible with all coupling methods.Regeneration of phosphotyrosine from the phosphodiamidate is effected by acid catalyzed hydrolysis using 90% aq. TFA.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS


Literature references

[1] H. G. Chao, et al. (1995) J. Org. Chem., 60, 7710.

Linkage

Replaces: 04-12-1205

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 96.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 93.0 %
Water (K. F.): ≤ 4.5 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service