Skip to Content
Merck
All Photos(1)

Key Documents

8.22000

Sigma-Aldrich

1-Chlorooctadecane

for synthesis

Synonym(s):

1-Chlorooctadecane, Stearyl chloride, Octadecyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17Cl
CAS Number:
Molecular Weight:
288.94
MDL number:
UNSPSC Code:
12352101
EC Index Number:
222-207-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

320 °C

mp

20-23 °C

transition temp

flash point 110 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCCCCCCCCCCCCCCCCCC

InChI

1S/C18H37Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3

InChI key

VUQPJRPDRDVQMN-UHFFFAOYSA-N

General description

1-Chlorooctadecane is a long-chain aliphatic compound that belongs to the class of alkyl halides. It is commonly used as a reagent in organic synthesis, particularly in the formation of alkyl chains in larger molecules. Its long hydrophobic alkyl chain makes it ideal for synthesis of surfactants, such as quaternary ammonium salts. The presence of the chlorine atom makes it susceptible to nucleophilic substitution reactions.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 20 °C
Melting range (upper value): ≤ 23 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service