Skip to Content
Merck
All Photos(1)

Key Documents

8.20557

Sigma-Aldrich

Isobutyl acetate

for synthesis

Synonym(s):

Isobutyl acetate, Acetic acid isobutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-745-1
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
116-118 °C/1013 hPa
vapor pressure:
17 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

17 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

510 °C

potency

13400 mg/kg LD50, oral (Rat)
>17400 mg/kg LD50, skin (Rabbit)

expl. lim.

2.4-10.5 % (v/v)

pH

5 (20 °C, 4 g/L in H2O)

bp

116-118 °C/1013 hPa

mp

-99 °C

transition temp

flash point 18 °C

solubility

7 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC(C)C)C(=O)C

InChI

1S/C6H12O2/c1-5(2)4-8-6(3)7/h5H,4H2,1-3H3

InChI key

GJRQTCIYDGXPES-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.870 - 0.872
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - Pensky-Martens closed cup

Flash Point(C)

22 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service