8.20151
di-Sodium succinate hexahydrate
for synthesis
Synonym(s):
di-Sodium succinate hexahydrate, Succinic acid disodium salt
About This Item
Recommended Products
Quality Level
form
crystals
pH
6.0-8.5 (20 °C, 50 g/L in H2O)
mp
120 °C (Elimination of water of crystallisation)
solubility
349 g/L
storage temp.
2-30°C
SMILES string
[Na+].[Na+].[O-]C(=O)CCC(=O)[O-].O.O.O.O.O.O
InChI
1S/C4H6O4.2Na.6H2O/c5-3(6)1-2-4(7)8;;;;;;;;/h1-2H2,(H,5,6)(H,7,8);;;6*1H2/q;2*+1;;;;;;/p-2
InChI key
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Application
- Application in nanoparticle synthesis: Di-Sodium succinate hexahydrate is employed in the robust synthesis of size-dispersed triangular silver nanoprisms, demonstrating a key role in the chemical reduction route. This approach contributes to the understanding of nanoparticle formation and could optimize the production processes for applications ranging from electronics to biomedicine (Bahlol et al., 2019).
Analysis Note
Loss on drying (130 °C, 2 h): 38.0 - 42.0 %
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service