Skip to Content
Merck
All Photos(1)

Key Documents

8.14114

Sigma-Aldrich

Saccharin sodium salt dihydrate

for synthesis

Synonym(s):

Saccharin sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4NNaO3S · 2H2O
CAS Number:
Molecular Weight:
241.20
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-886-1
NACRES:
NA.22

Quality Level

potency

14200 mg/kg LD50, oral (Rat)

pH

6.0-7.5 (20 °C, 100 g/L in H2O)

transition temp

flash point >200 °C

solubility

1000 g/L

bulk density

600‑800 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[S]1(=NC(=O)c2c1cccc2)(=O)[O-].O.O

InChI

1S/C7H5NO3S.Na.2H2O/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;;;/h1-4H,(H,8,9,10,11);;2*1H2/q;+1;;/p-1

InChI key

NAUHOSQQKJTUAX-UHFFFAOYSA-M

Application

  • Saccharin Anion in Zn2+ Electrochemistry: Investigates the role of saccharin sodium in enhancing the performance of zinc anodes in batteries, highlighting its potential in electrolyte modification for better energy storage solutions (Xi et al., 2021).
  • Behavioral Responses to Saccharin Mixtures in Chicks: Examines the preferences of chicks for saccharin mixed with quinine, contributing to sensory science and flavor enhancement research (Yoshida et al., 2018).

Analysis Note

Assay (HClO₄, calc. on anhydrous substance): ≥ 99.0 %
Water (K. F.): 13.0 - 15.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service