Skip to Content
Merck
All Photos(1)

Key Documents

8.08076

Sigma-Aldrich

Thioacetic acid

for synthesis

Synonym(s):

Thioacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4OS
CAS Number:
Molecular Weight:
76.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-063-8
NACRES:
NA.22

grade

for synthesis

Quality Level

vapor pressure

1 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

potency

200-350 mg/kg LD50, oral (Rat)

pH

1.8 (20 °C, 100 g/L in H2O, Hydrolysis)

mp

-17 °C

transition temp

flash point 18 °C

solubility

27 g/L (slow decomposition)

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SC(=O)C

InChI

1S/C2H4OS/c1-2(3)4/h1H3,(H,3,4)

InChI key

DUYAAUVXQSMXQP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.067 - 1.070
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service