Skip to Content
Merck
All Photos(1)

Key Documents

8.00396

Sigma-Aldrich

Hexacarbonylmolybdenum

for synthesis

Synonym(s):

Hexacarbonylmolybdenum, Molybdenum carbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mo(CO)6
CAS Number:
Molecular Weight:
264.00
MDL number:
UNSPSC Code:
12161603
EC Index Number:
237-713-3
NACRES:
NA.22

vapor pressure

0.13 hPa ( 20 °C)

Quality Level

form

powder

mp

147-152 °C (decomposition)

density

1.96 g/cm3 at 20 °C

SMILES string

[Mo].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C]

InChI

1S/6CO.Mo/c6*1-2;

InChI key

KMKBZNSIJQWHJA-UHFFFAOYSA-N

Analysis Note

Assay (Mo(CO)₆): ≥ 99.0 %
Assay (Cr(CO)₆): ≤ 1.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service