Skip to Content
Merck
All Photos(1)

Key Documents

616420

Sigma-Aldrich

Transferrin, Holo, Bovine Plasma

Synonym(s):

Transferrin, Holo, Bovine Plasma

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 9571

Quality Level

Assay

≥95% (FPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤30 EU/mg Endotoxins (LAL test)

color

red-brown

solubility

water: 1% (Readily soluble)
20 mM Tris-HCl buffer, pH 7.5: soluble

cation traces

Fe: ≥1000 ppm

shipped in

ambient

storage temp.

−20°C

General description

Native holo-transferrin from bovine plasma. Transferrin is a major iron binding protein and is found in human plasma or serum at concentrations of 3 mg/ml. Its concentration correlates with the total iron binding capacity of serum and its level is regulated by the availability of iron. It has bacteriostatic and fungistatic activity and is essential for iron utilization by the hemin-synthesizing reticulocytes. Stored in bone marrow as Tf-bound iron.

Warning

Toxicity: Standard Handling (A)

Physical form

Salt-free.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or add NaN₃ and refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 1 year at -20°C or for up to 2 weeks at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service