Skip to Content
Merck
All Photos(1)

Key Documents

580217

Millipore

Taurocholic Acid, Sodium Salt

Detergent useful for the solubilization of lipids and membrane-bound proteins. Aggregation number: 4.

Synonym(s):

Taurocholic Acid, Sodium Salt, Sodium Taurocholate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NO7S · Na
CAS Number:
Molecular Weight:
537.68
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

Quality Level

description

Merck USA index - 14, 9075

Assay

≥90% (TLC)

form

solid

mol wt

micellar wt 2100

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

aggregation number

4

color

white

pKa 

1.9

CMC

3 - 11 mM

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCNC(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](C[C@H]3O)C[C@@H](CC4)O)C)C[C@@H]2O)CC1)C)C

InChI

1S/C26H45NO7S.Na/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,22+,24+,25+,26-;/m1./s1

InChI key

JAJWGJBVLPIOOH-IZYKLYLVSA-M

General description

Detergent useful for the solubilization of lipids and membrane-bound proteins (aggregation number: 4; CMC: 3-11 mM; micellar weight: 2100; pKa: 1.9).

Application


  • Discovery of Taurocholic Acid Sodium Hydrate as a Novel Repurposing Drug for Intervertebral Disc Degeneration by Targeting MAPK3.: This research highlights the therapeutic potential of taurocholic acid, sodium salt in treating intervertebral disc degeneration. By targeting the MAPK3 pathway, it offers a new approach to managing spinal disorders, reflecting its significance in developing innovative treatments (Li P et al., 2024).

  • Effects of extraction methods and particle size distribution on the structural, physicochemical, and functional properties of dietary fiber from deoiled cumin.: This research uses taurocholic acid, sodium salt to investigate the optimization of dietary fiber extraction, contributing to advancements in food chemistry and health-related product development (Ma MM et al., 2016).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service