Skip to Content
Merck
All Photos(1)

Key Documents

577801

Sigma-Aldrich

Anti-Tau Mouse mAb (TAU-5)

liquid, clone TAU-5, Calbiochem®

Synonym(s):

Anti-Tau antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

TAU-5, monoclonal

form

liquid

does not contain

preservative

species reactivity

mouse, human, sheep, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

General description

Protein G purified mouse monoclonal antibody. Recognizes the ~45-68 kDa phosphorylated and unphosphorylated forms of tau.
Recognizes the ~45-68 kDa Tau protein.
This Anti-Tau Mouse mAb (TAU-5) is validated for use in Frozen Sections, Immunoblotting, Immunoprecipitation, Paraffin Sections for the detection of Tau.

Immunogen

Bovine
Epitope: within the central region
purified bovine microtubule-associated proteins

Application

Frozen Sections (1-2 µg/ml)

Immunoblotting (1 µg/ml)

Immunoprecipitation (10 µg per 200-500 µg cell lysate)

Paraffin Sections (1-2 µg/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Do not store diluted antibody without carrier protein if the concentration is <50 µg/ml.

Other Notes

Papasozomenos, S.C. and Shanavas, A., 2002. Proc. Natl. Acad. Sci. USA99, 1140.
Rapoport, M., eta al. 2002. Proc. Natl. Acad. Sci. USA99, 6364.
When used for formalin/paraffin embedded sections, staining is enhanced by boiling tissue sections in 10 mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at room temperature for 20 min prior to antibody incubation. Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau. Does not cross-react with other MAPS of tubulin. Recognizes both native and phosphorylated forms of tau. Variables associated with assay conditions will dictate proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ricardo Gargini et al.
Frontiers in aging neuroscience, 11, 231-231 (2019-09-26)
The analysis of global and comparative genomics between different diseases allows us to understand the key biological processes that explain the etiology of these pathologies. We have used this type of approach to evaluate the expression of several neurodegeneration-related genes
Michael Dumbacher et al.
Molecular neurodegeneration, 13(1), 50-50 (2018-09-28)
Neuronal Ca2+ dyshomeostasis and hyperactivity play a central role in Alzheimer's disease pathology and progression. Amyloid-beta together with non-genetic risk-factors of Alzheimer's disease contributes to increased Ca2+ influx and aberrant neuronal activity, which accelerates neurodegeneration in a feed-forward fashion. As
Weijiang Dong et al.
Journal of neuroscience research, 87(14), 3176-3185 (2009-05-28)
Tau function is regulated by phosphorylation, and abnormal tau phosphorylation in neurons is one of the key processes associated with development of Alzheimer's disease and other tauopathies. In this study we provide evidence that phospholipid transfer protein (PLTP), one of
Ram Fridlich et al.
Molecular & cellular proteomics : MCP, 8(6), 1206-1218 (2009-03-13)
Rod-derived cone viability factor (RdCVF) is produced by the Nxnl1 gene that codes for a second polypeptide, RdCVFL, by alternative splicing. Although the role of RdCVF in promoting cone survival has been described, the implication of RdCVFL, a putative thioredoxin
Marta Fernández-Nogales et al.
Brain pathology (Zurich, Switzerland), 27(3), 314-322 (2016-06-25)
Increased incidence of neuronal nuclear indentations is a well-known feature of the striatum of Huntington's disease (HD) brains and, in Alzheimer's disease (AD), neuronal nuclear indentations have recently been reported to correlate with neurotoxicity caused by improper cytoskeletal/nucleoskeletal coupling. Initial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service