Skip to Content
Merck
All Photos(1)

Key Documents

557370

Sigma-Aldrich

Rottlerin

≥98% (HPLC), solid, PKC inhibitor, Calbiochem®

Synonym(s):

Rottlerin, Mallotoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28O8
CAS Number:
Molecular Weight:
516.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Rottlerin, A cell-permeable and reversible protein kinase C inhibitor that exhibits greater selectivity for PKCδ (IC₅₀ = 3-6 µM) and PKCθ.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange

solubility

ethanol: 1 mg/mL
DMSO: 30 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1C(C=Cc2c1c(c(c(c2O)Cc4c(c(c(c(c4O)C(=O)C)O)C)O)O)C(=O)\C=C\c3ccccc3)(C)C

InChI

1S/C30H28O8/c1-15-24(33)19(27(36)22(16(2)31)25(15)34)14-20-26(35)18-12-13-30(3,4)38-29(18)23(28(20)37)21(32)11-10-17-8-6-5-7-9-17/h5-13,33-37H,14H2,1-4H3/b11-10+

InChI key

DEZFNHCVIZBHBI-ZHACJKMWSA-N

General description

A cell-permeable and reversible inhibitor of protein kinase C δ (PKCδ, IC50 = 3-6 µM) and PKCΘ isozymes. Also inhibits PKCα, PKCβ, and PKCγ isoforms, but with significantly reduced potency (IC50 = 30-42 µM). Has reduced inhibitory activity on PKCε, PKCη, and PKCξ (IC50 = 80-100 µM). Also known to inhibit CaM kinase III (IC50 = 5.3 µM).
A cell-permeable and reversible protein kinase C inhibitor that exhibits greater selectivity for PKCδ (IC50 = 3-6 µM) and PKCθ. Inhibits the PKCα, PKCβ, and PKCγ isoforms, but with significantly reduced potency (IC50 = 30-42 µM). Has reduced inhibitory activity on PKCε, PKCη, and PKCζ (IC50 = 80-100 µM). Also known to inhibit CaM kinase III (IC50 = 5.3 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKCδ
Product does not compete with ATP.
Reversible: yes
Target IC50: 3-6 µM against PKCδ; 5.3 µM against CaM kinase III

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Villalba, M., et al. 1999. J. Immunol. 163, 5813.
Gschwendt, M., et al. 1994. Biochem. Biophys. Res. Commun. 199, 93.
Gschwendt, M., et al. 1994. FEBS Lett.338, 85.
Gschwendt, M., et al. 1984. Cancer Lett.25, 177.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Helios Gallego-Gutiérrez et al.
Molecular biology of the cell, 32(15), 1347-1358 (2021-05-20)
The presence of tight junction protein zonula occludens 2 (ZO-2) at the nucleus inhibits the transcription of genes regulated by TEAD transcription factor. Here, we analyzed whether the movement of ZO-2 into the nucleus modulates the nuclear concentration of TEAD.
Hyeong-Jae Kim et al.
International journal of molecular sciences, 23(16) (2022-08-27)
Lamins are nuclear envelope proteins involved in various cellular functions, such as DNA modulation, cellular differentiation, and development. In this study, we investigate the role of histamine in lung cancer biology. Since it is known that lamin-A/C is negatively regulated
Yan-Wei Wu et al.
Materials today. Bio, 17, 100480-100480 (2022-11-11)
Most existing vaccines use activators that polarize the immune response to T-helper (Th) 2 response for antibody production. Our positively charged chitosan (Cs)-based nanocomplex (CNC) drives the Th1 response through unknown mechanisms. As receptors for the positively charged CNC are
Shahnawaz A Bhat et al.
Journal of neurochemistry, 156(2), 225-248 (2020-01-12)
We have previously shown that treatment with a mGluR5 positive allosteric modulator (PAM) is neuroprotective after experimental traumatic brain injury (TBI), limiting post-traumatic neuroinflammation by reducing pro-inflammatory microglial activation and promoting anti-inflammatory and neuroprotective responses. However, the specific molecular mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service