Skip to Content
Merck
All Photos(2)

Key Documents

527450

Sigma-Aldrich

Imidazolo-oxindole PKR inhibitor C16

≥90% (HPLC), solid, PKR inhibitor, Calbiochem®

Synonym(s):

PKR Inhibitor, Double-stranded RNA-dependent Protein Kinase Inhibitor, C16, Double-stranded RNA-activated Protein Kinase Inhibitor I, EIF2AK2 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N4OS
CAS Number:
Molecular Weight:
268.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

PKR Inhibitor, The PKR Inhibitor, also referenced under CAS 608512-97-6, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange to orange-brown

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c(nc1)ccc3c2\C(=C\c4nc[nH]c4)\C(=O)N3

InChI

1S/C13H8N4OS/c18-13-8(3-7-4-14-5-15-7)11-9(17-13)1-2-10-12(11)19-6-16-10/h1-6H,(H,14,15)(H,17,18)/b8-3-

InChI key

VFBGXTUGODTSPK-BAQGIRSFSA-N

General description

An imidazolo-oxindole compound that acts as a potent, ATP-binding site directed inhibitor of PKR. Shown to effectively inhibit RNA-induced PKR autophosphorylation (IC50 = 210 nM) and rescue PKR-dependent translation block (IC50 = 100 nM). Inactive control is also available. Shown to increase the late phase of long-lasting synaptic potentiation, and enhance long-term memory in mice. Also available as a 50 mM solution in DMSO (Cat. No. 527451).

Biochem/physiol Actions

Cell permeable: no
Primary Target
PKR
Product does not compete with ATP.
Reversible: no
Target IC50: 210 nM inhibiting RNA-induced PKR autophosphorylation and 100 nM in rescuing PKR-dependent translation block

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhu, P.J., et al. 2011. Cell147, 1384.
Jammi, N.V., et al. 2003. Biochem. Biophys. Res. Commun.308, 50.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lasse Reimer et al.
Brain pathology (Zurich, Switzerland), 31(1), 103-119 (2020-07-28)
Deposition of extensively hyperphosphorylated tau in specific brain cells is a clear pathological hallmark in Alzheimer's disease and a number of other neurodegenerative disorders, collectively termed the tauopathies. Furthermore, hyperphosphorylation of tau prevents it from fulfilling its physiological role as
Kenneth T Farabaugh et al.
eLife, 9 (2020-03-17)
The inability of cells to adapt to increased environmental tonicity can lead to inflammatory gene expression and pathogenesis. The Rel family of transcription factors TonEBP and NF-κB p65 play critical roles in the switch from osmoadaptive homeostasis to inflammation, respectively.
Sanket S Ponia et al.
Cell reports, 37(4), 109888-109888 (2021-10-28)
Dysregulated inflammation dominated by chemokine expression is a key feature of disease following infection with the globally important human pathogens Zika virus (ZIKV) and dengue virus, but a mechanistic understanding of how pro-inflammatory responses are initiated is lacking. Mitophagy is
Upayan Patra et al.
Oxidative medicine and cellular longevity, 2020, 7289120-7289120 (2020-04-24)
Eukaryotic cells adopt highly tuned stress response physiology under threats of exogenous stressors including viruses to maintain cellular homeostasis. Not surprisingly, avoidance of cellular stress response pathways is an essential facet of virus-induced obligatory host reprogramming to invoke a cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service