Skip to Content
Merck
All Photos(1)

Key Documents

264101

Millipore

Deoxycholic Acid sodium

An ionic detergent for the solubilization of membrane-bound proteins.

Synonym(s):

Sodium deoxycholate, 3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO4
CAS Number:
Molecular Weight:
414.55
Beilstein:
3581950
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 2899

Assay

≥96% (TLC)

form

powder

mol wt

micellar avg mol wt 1200-5000
micellar wt 1700-4200

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

aggregation number

3-12
4 - 10

color

white to off-white

pKa 

6.2

CMC

2-6 mM (20-25°C)
4 - 8 mM

solubility

H2O: 2%, clear, colorless

cation traces

heavy metals: ≤20.0 ppm

HLB

16

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC([O-])=O)[C@@]4(C)[C@@H](O)C[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C24H40O4.Na/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;/m1./s1

InChI key

FHHPUSMSKHSNKW-SMOYURAASA-M

Looking for similar products? Visit Product Comparison Guide

General description

An ionic detergent for the solubilization of membrane-bound proteins (aggregation number: 4-10; CMC: 4-8 mM; micellar weight: 1700-4200; pKa 6.2). Reported to induce tyrosine phosphorylation of β-catenin and its subsequent translocation to the nucleus, promoting activation of β-catenin signaling (effective concentration 5-50 µM).

Warning

Toxicity: Harmful (C)

Other Notes

Pai, R., et al. 2004. Mol. Biol. Cell15 2156.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service