Skip to Content
Merck
All Photos(1)

Key Documents

182200

Sigma-Aldrich

βARK1 Inhibitor

The βARK1 Inhibitor, also referenced under CAS 24269-96-3, controls the biological activity of βARK1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

βARK1 Inhibitor, β-Adrenergic Receptor Kinase1 Inhibitor, GRK2 Inhibitor, Methyl 5-[2-(5-nitro-2-furyl)vinyl]-2-furoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NO6
CAS Number:
Molecular Weight:
263.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to orange

solubility

DMSO: 5 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1[o]c(cc1)\C=C\c2[o]c(cc2)C(=O)OC

InChI

1S/C12H9NO6/c1-17-12(14)10-6-4-8(18-10)2-3-9-5-7-11(19-9)13(15)16/h2-7H,1H3/b3-2+

InChI key

YDJPHSNZGRVPCK-NSCUHMNNSA-N

General description

A selective inhibitor of β-adrenergic receptor kinase 1 (βARK1) (IC50 = 126 µM). Displays no inhibitory activity against protein kinase A (PKA). Identified from a group of existing compounds (not previously demonstrated to inhibit βARK1) using a two-step rational drug design protocol that included molecular modeling and comparative docking studies. Represents the first inhibitor that distinguishes between βARK1 and PKA, an enzyme with a similar adenine binding pocket. May be useful in the study of mechanisms involved in congestive heart failure where elevated levels of βARK1 desensitize agonist-bound β2-adrenergic receptors.
A selective inhibitor of βARK1 (β-Adrenergic Receptor Kinase 1) (IC50 = 126 µM). Has no inhibitory activity against protein kinase A (PKA).

Biochem/physiol Actions

Cell permeable: no
Primary Target
βARK1 (β-Adrenergic Receptor Kinase 1)
Product does not compete with ATP.
Reversible: no
Target IC50: 126 µM against βARK1 (β-Adrenergic Receptor Kinase 1)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Iino, M., et al. 2002. J. Med. Chem.45, 2150.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service