Skip to Content
Merck
All Photos(1)

Key Documents

178481

Sigma-Aldrich

Transferrin, Apo-, Low Endotoxin Grade, Human Plasma

Synonym(s):

Transferrin, Apo-, Low Endotoxin Grade, Human Plasma

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 9571

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<0.1 EU/mg Endotoxins

color

white

pI 

5.8

solubility

water: 10 mg/mL

cation traces

Fe: ≤0.01%

shipped in

ambient

storage temp.

−20°C

General description

Native apotransferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free. Cell culture tested.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 10 mM phosphate buffer, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service