Skip to Content
Merck
All Photos(1)

Key Documents

17-283

Sigma-Aldrich

Rac1 Activation Assay Kit

Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Rac1-GTP from cell lysates & detection by a Rac1 specific monoclonal antibody.

Synonym(s):

Activation Assay Kit, Rac1 Activity Kit, Rac1 Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Quality Level

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable
affinity binding assay: suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... RAC1(5879)

General description

Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Ras-related C3 botulinum toxin substrate 1 (Rac1)-GTP from cell lysates and detected by a Rac1 specific monoclonal antibody. Rac1 belongs to the Ras superfamily of small guanosine triphosphatases (GTPases). It is expressed in two major forms such as an inactive GDP-bound form and an active GTP-bound form. Rac1 is involved in influencing several physiological conditions by interacting with effectors that activate several signaling cascades. The RAC1 gene is located on the human chromosome at 7p22.1.

Application

Rac1 Activation Assay Kit has been used in the detection of active Ras-related C3 botulinum toxin substrate 1 (Rac1) in brain homogenates of mice and humans by immunoblotting.

Components

Anti-Rac1, clone 23A8 (Cat.# 05-389)

Mg2+ Lysis/Wash Buffer, 5X (Cat.# 20-168)

100X GTPγS, 10mM (Cat.# 20-176)

100X GDP, 100mM (Cat.# 20-177)

Rac/cdc42 Assay Reagent (PAK1 PBD, agarose) (Cat.# 14-325)

Quality

Routinely evaluated by precipitating Rac-GTP from 3T3 cell lysates that had been loaded with GTPγS. The precipitated Rac-GTP was detected by immunoblot analysis using anti-Rac.

Linkage

Replaces: SGT425

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of mechanical stretching in the activation and localization of adhesion proteins and related intracellular molecules.
Boccafoschi, F, et al.
Journal of Cellular Biochemistry, 112, 1403-1409 (2011)
The survival kinase Mirk/dyrk1B is activated through Rac1-MKK3 signaling.
Kideok Jin, Seunghwan Lim, Stephen E Mercer, Eileen Friedman
The Journal of Biological Chemistry null
rs12977 of RAC1 is associated with the progress of gastric cancer in Iranian population
Javani A, et al.
Gene Reports, 23, 101069-101069 (2021)
Incorporation of CXCR4 into membrane lipid rafts primes homing-related responses of hematopoietic stem/progenitor cells to an SDF-1 gradient.
Marcin Wysoczynski, Ryan Reca, Janina Ratajczak, Magda Kucia, Neeta Shirvaikar et al.
Blood null
Anti-arrhythmic surgery for atrioventricular junctional and atrioventricular re-entrant tachycardia--a report of six cases.
Koo, C C, et al.
Annals of the Academy of Medicine, Singapore, 19, 67-72 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service