Skip to Content
Merck
All Photos(1)

Key Documents

1.70328

Supelco

Lead ICP standard

traceable to SRM from NIST Pb(NO₃)₂ in HNO₃ 2-3% 1000 mg/l Pb Certipur®

Synonym(s):

Nota 1 A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

product line

Certipur®

technique(s)

ICP: suitable

pH

0.5 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

  • Environmental Contamination Analysis:The study utilized ICP-MS, employing Lead ICP standards to analyze toxic elements in soil around a waste management site. This research underscores the crucial role of Lead ICP standards in tracing and quantifying environmental contaminants (Olanipekun OO et al., 2024).
  • Calibration of Analytical Instruments: In a study microwave-induced combustion was used for the determination of toxic elements in honey samples by ICP-MS, where Lead ICP standards were essential for ensuring the accuracy and reliability of measurements, highlighting their importance in food safety and biochemical analysis (Cauduro VH et al., 2024).
  • Development of Electrochemical Sensors:The study developed screen-printed sensors modified with Nafion and mesoporous carbon for the electrochemical detection of lead in blood. Lead ICP standards played a key role in calibrating these sensors, demonstrating their utility in medical and biochemical sensor development (Boselli E et al., 2024).

Other Notes

Blocked in the US and Canada due to regulatory reasons
For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.

A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.

A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.

A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service