Skip to Content
Merck
All Photos(1)

Key Documents

1.59267

Supelco

Metanil yellow

pH indicator, Reag. Ph Eur

Synonym(s):

Metanil yellow, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N3NaO3S
CAS Number:
Molecular Weight:
375.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-608-2
NACRES:
NA.21

Product Name

Metanil yellow, Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

color

brownish-yellow

pH

10 (20 °C, 10 g/L in H2O)

solubility

25 g/L

bulk density

330 kg/m3

storage temp.

no temp limit

SMILES string

[Na+].[S](=O)(=O)([O-])c1cc(ccc1)N=Nc2ccc(cc2)Nc3ccccc3

InChI

1S/C18H15N3O3S.Na/c22-25(23,24)18-8-4-7-17(13-18)21-20-16-11-9-15(10-12-16)19-14-5-2-1-3-6-14;/h1-13,19H,(H,22,23,24);/q;+1/p-1

InChI key

NYGZLYXAPMMJTE-UHFFFAOYSA-M

Analysis Note

Description: brownish-yellow powder
Transition range: pH 1.2 - 2.3
Sensitivity test: conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service