1.37011
Acetic acid
75%, EMPROVE® EXPERT
Pharma Manufacturing
Synonym(s):
Acetic acid, Essigsäure, Glacial acetic acid
About This Item
Recommended Products
grade
GMO free
GMP
Halal
Kosher
Quality Level
vapor density
2.07 (vs air)
product line
EMPROVE® EXPERT
refractive index
n20/D 1.371 (lit.)
bp
117-118 °C (lit.)
application(s)
cleaning in place
pharma/biopharma processes
pharmaceutical
foreign activity
Aflatoxin free
SMILES string
[F2C(F2C)13F3C]C(O)=O
InChI
1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
InChI key
QTBSBXVTEAMEQO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. OurSAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Packaging
1.37011.9025 // 25L // plastic container
1.37011.9950 // 950L // PE-drum
1.37011.9200 // 200L // PE-drum
Legal Information
also commonly purchased with this product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service