Skip to Content
Merck
All Photos(1)

Key Documents

12659-M

Sigma-Aldrich

Albumin, Bovine Serum, Fraction V, Low Heavy Metals

Synonym(s):

Albumin, Bovine Serum, Fraction V, Low Heavy Metals

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 8468

Assay

≥98% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤10% moisture
<2.0% Ash

color

beige

solubility

aqueous buffer: 10 mg/mL
water: 10 mg/mL

cation traces

heavy metals: ≤20 ppm

storage temp.

2-8°C

General description

Prepared by heat shock treatment procedure. Useful for the preparation of protein and molecular weight standards, and as a protein base for sensitive diagnostic and research assays, such as RIA, ELISA, and vitamin assay. Material can also be used as an antigen carrier protein in immunological studies and monoclonal antibody production, and as a control protein for enzymatic assays, such as protease, perioxidase, phosphatase, or nuclease when determining the purity of reagent grade albumins.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). 0.1% NaN₃ may be added as a preservative. Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Cohn, E.J., et al. 1947. J. Am. Chem. Soc.69, 1753.
Cohn, E.J., et al. 1946. J. Am. Chem. Soc.68, 1946.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel D Haschka et al.
Cell death and differentiation, 27(8), 2297-2312 (2020-02-06)
Cells experiencing delays in mitotic progression are prone to undergo apoptosis unless they can exit mitosis before proapoptotic factors reach a critical threshold. Microtubule targeting agents (MTAs) arrest cells in mitosis and induce apoptotic cell death engaging the BCL2 network.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service