1.09948
Chromium standard
1000 mg Cr, (CrCl₃ in 4.2% HCl) Titrisol®
Synonym(s):
Chromium Calibration Standard for ICP/AAS
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
Titrisol®
form
concentrate
concentration
1000 mg Cr in 4.2% HCl
technique(s)
AAS: suitable
ICP: suitable
pH
<1 (20 °C in H2O)
density
1.05 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
single component solution
storage temp.
15-25°C
Features and Benefits
Analysis Note
Determination method: ICP - OES.
(traceable to NIST - SRM 3112a)
Accuracy of the method: +/- 5 mg
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Met. Corr. 1 - Skin Sens. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service