Skip to Content
Merck
All Photos(1)

Key Documents

1.09637

Supelco

Methyl decanoate

reference substance for gas chromatography

Synonym(s):

Methyl decanoate, Methyl caprate, Capric acid methyl ester, Decanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22O2
CAS Number:
Molecular Weight:
186.29
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-766-6
NACRES:
NA.24

Quality Level

form

liquid

expl. lim.

0.7 % (v/v)

bp

>176.7 °C/1013 hPa

mp

-18--10 °C

transition temp

flash point 90 °C

solubility

0.0038 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCCCCCCC

InChI

1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3

InChI key

YRHYCMZPEVDGFQ-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99.5
Identity (IR-spectrum): conforms

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.9 °F - Pensky-Martens closed cup

Flash Point(C)

110.5 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service