1.09437
Buffer solution
(citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 6.00 (20°C) Certipur®
Synonym(s):
Buffer solution pH 6.00 (20°C) (citric acid/sodium hydroxide)
About This Item
Recommended Products
grade
certified reference material
Quality Level
product line
Certipur®
pH
6.0 (20 °C in H2O)
density
1.01 g/cm3 at 20 °C
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
Related Categories
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Ready-to-use solution subjected to comprehensive stability studies in accordance with ISO Guide 35
- Available in a sealed bottle and the innovative Titripac® packaging for positive environmental impact due to reduced waste generation
- Long-lasting with a shelf life of 3 years
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to SRMs from NIST and primary certified reference standards from PTB
Legal Information
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service