1.06577
tri-Sodium phosphate dodecahydrate
EMPROVE® ESSENTIAL, NF, E 339
Pharma Manufacturing
Synonym(s):
tri-Sodium phosphate dodecahydrate, Sodium phosphate tribasic, Trisodium phosphate
About This Item
Agency
NF
Quality Level
product line
EMPROVE® ESSENTIAL
form
solid
pH
12 (20 °C, 10 g/L in H2O)
mp
75 °C (Elimination of water of crystallisation)
solubility
285 g/L
density
1.62 g/cm3 at 20 °C
application(s)
liquid formulation
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
15-25°C
SMILES string
[Na+].[Na+].[Na+].[P](=O)([O-])([O-])[O-].O.O.O.O.O.O.O.O.O.O.O.O
InChI
1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3
InChI key
ASTWEMOBIXQPPV-UHFFFAOYSA-K
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service