Skip to Content
Merck
All Photos(5)

Key Documents

1.02832

Supelco

Cyclohexane

EMPLURA®

Synonym(s):

Hexahydrobenzene, Hexamethylene, Naphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-806-2
NACRES:
NA.21
Assay:
≥99% (GC)
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Quality Level

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

500 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

9 %

impurities

≤0.001% Free acid (as CH3COOH)
≤0.05% Water

refractive index

n20/D 1.426 (lit.)

kinematic viscosity

1.26 cSt(20 °C)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

flash point -18 °C

solubility

0.05 g/L

density

0.779 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Application

Cyclohexane can be used as a solvent:
  • To study thermal degradation of low-density polyethylene (LDPE) wastes.[1]
  • In the enantioselective reductive alkylation reaction in the presence of Ir-ferrocenyl diphosphine as a catalyst.[2]
  • To synthesize hydroxyl-terminated polybutadiene (HTPB) via anionic polymerization of butadiene in the presence of an initiator.[3]
  • In the enantioselective hydrogenation of ethyl pyruvate to R(+)ethyl lactate in the presence of Pt/zeolite catalyst.[4]

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.001 %
Density (d 20 °C/ 4 °C): 0.778 - 0.779
Readily carbonizable substances: conforms
Water: ≤ 0.05 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enantioselective hydrogenation of ethyl pyruvate: Long-term performance of chirally modified Pt/zeolite catalysts
Bohmer U, et al.
Catalysis Today, 60(3-4) (2000)
Synthesis of hydroxyl-terminated polybutadiene possessing high content of 1, 4-units via anionic polymerization
Chen Ji-ming, et al.
Chinese Journal of Polymer Science, 28(5) (2010)
Use of cyclohexane as solvent in thermal degradation of low density polyethylene wastes
Karaduman A, et al.
Polymer-Plastics Technology and Engineering, 41(4) (2002)
Iridium ferrocenyl diphosphine catalyzed enantioselective reductive alkylation of a hindered aniline
Blaser H-U, et al.
Synlett, 1999(1) (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service