Skip to Content
Merck
All Photos(1)

Key Documents

1.02766

Supelco

Copper(II) oxide

powder for analysis EMSURE® ACS

Synonym(s):

Copper(II) oxide, Copper monoxide granulated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-269-1
NACRES:
NA.21
grade:
ACS reagent
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

potency

470 mg/kg LD50, oral (Rat)

impurities

≤0.01% Total sulfur (as SO4)
≤0.02% Substances insoluble in dilute hydrochloric acid

particle size

≤160 μm (d95,laser diffraction)

pH

7 (20 °C, 50 g/L in H2O, slurry)

mp

1326 °C (decomposition)

density

6.48 g/cm3 at 25 °C

bulk density

500 kg/m3

anion traces

chloride (Cl-): ≤0.005%
sulfur compounds (as SO42-): ≤0.01%

cation traces

C: ≤0.01%
Ca: ≤0.01%
Cr: ≤0.05%
Fe: ≤0.05%
K: ≤0.02%
N: ≤0.002%
Na: ≤0.05%
Pb: ≤0.05%
excess alkali:, passes test

storage temp.

no temp limit

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Related Categories

Application


  • Impact of temperature on the physicochemical, structural and biological features of copper-silica nanocomposites.: This study investigates how temperature variations affect the properties of copper-silica nanocomposites. The research focuses on the physicochemical and structural changes that occur and their subsequent impact on biological applications. The findings provide valuable insights for the development of advanced materials with tailored properties for specific applications (Dulski et al., 2020).

  • H(2)O(2)-assisted photoelectrocatalytic degradation of Mitoxantrone using CuO nanostructured films: Identification of by-products and toxicity.: This article explores the degradation of Mitoxantrone, a chemotherapy drug, using copper oxide nanostructured films in a photoelectrocatalytic process assisted by hydrogen peroxide. The study identifies the by-products formed during degradation and assesses their toxicity. The research highlights the potential of CuO nanostructured films in environmental remediation and pharmaceutical waste management (da Rosa et al., 2019).

Analysis Note

Assay (complexometric): ≥ 99.0 %
Substances insoluble in dilute hydrochloric acid: ≤ 0.02 %
Free Alkali: passes test
Chloride (Cl): ≤ 0.005 %
Total sulfur (as SO₄): ≤ 0.01 %
Total nitrogen (N): ≤ 0.002 %
C (Carbon): ≤ 0.01 %
Ca (Calcium): ≤ 0.01 %
Cr (Chromium): ≤ 0.05 %
Fe (Iron): ≤ 0.05 %
K (Potassium): ≤ 0.02 %
Na (Sodium): ≤ 0.05 %
Pb (Lead): ≤ 0.05 %
Particle size (laser diffraction) (d95): ≤ 160 µm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service