Skip to Content
Merck
All Photos(1)

Key Documents

1.00615

Supelco

Potassium Test, photometric

photometric, 30-300 mg/L (K), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Potassium Cell Test, photometric, 30-300 mg/L (K), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

potassium

measuring range

30-300 mg/L (K)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Kalignost®, turbidimetric)

storage temp.

15-25°C

General description

Method: photometric 30 - 300 mg/l K Spectroquant®
This Spectroquant® Potassium Cell Test allows the accurate quantification of the potassium content in aqueous samples.
Method applied: In alkaline solution potassium ions react with Kalignost® (sodium tetraphenylborate) to form a slightly soluble precipitate. The resulting turbidity is measured in the photometer (turbidimetric method).The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

Kalignost is a registered trademark of Heyl Chemisch-Pharmazeutische Fabrik GmbH & Co. KG
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service