Skip to Content
Merck
All Photos(1)

Key Documents

07-685

Sigma-Aldrich

Anti-Lactotransferrin Antibody

unpurified, Upstate®

Synonym(s):

Anti-Draculin, Anti-Draculin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IHC (p)
IP
WB
species reactivity:
hamster, human, monkey, rat, mouse
technique(s):
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
17

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

hamster, human, monkey, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LTF(4057)

Specificity

Lactotransferrin

Immunogen

Full length native mouse Lactotransferrin/Lactoferrin

Application

Anti-Lactotransferrin Antibody is an antibody against Lactotransferrin for use in IP, WB, IC, IH, IH(P).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Routinely evaluated by immunoblot in mouse uterine fluid.

Target description

76-80kda

Physical form

100µl of rabbit antiserum, 0.05% sodium azide

Storage and Stability

2 years at 2-8°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Technology evaluation: rh lactoferrin, Agennix.
Andersen, Jeanette H
Current Opinion in Molecular Therapeutics, 6, 344-349 (2004)
K Nowicka-Bauer et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 69(3) (2018-08-28)
Reduced sperm motility, defined as asthenozoospermia, is a frequent cause of male infertility, and is mainly connected with the dysfunction of sperm mitochondria. The aim of this study was to identify the proteins, and thereby the metabolic pathways, responsible for
Lactoferrin functions: current status and perspectives.
Valenti, Piera, et al.
Journal of Clinical Gastroenterology, 38, S127-S129 (2004)
Lactoferrin gene expression and regulation: an overview.
Teng, Christina T
Biochemistry and Cell Biology = Biochimie Et Biologie Cellulaire, 80, 7-16 (2002)
Quantification of the host response proteome after mammalian reovirus T1L infection.
Berard, AR; Cortens, JP; Krokhin, O; Wilkins, JA; Severini, A; Coombs, KM
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service