Skip to Content
Merck
All Photos(3)

Key Documents

05-1471

Sigma-Aldrich

Anti-Rictor Antibody, clone 9F1.2

clone 9F1.2, from mouse

Synonym(s):

Rictor, AVO3, rapamycin-insensitive companion of mTOR, mAVO3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F1.2, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RICTOR(253260)

General description

Rictor, the rapamycin-insensitive complex, is part of the mTORC2 kinase complex which is responsible for phosphorylating the hydrophobic motif kinase of Akt/PKB. This complex also includes mTOR, mSin1 and mLST8/GL. mTORC2 is activated by growth factors and other mitogenic signals.

Specificity

This antibody recognizes Rictor.

Immunogen

Recombinant protein corresponding to human Rictor.

Application

Anti-Rictor Antibody, clone 9F1.2 detects level of Rictor & has been published & validated for use in WB & IP.
Immunoprecipitation:
Representative lot data.
1 ug/mL dilution of this antibody immunoprecipitated Rictor from 100 mg of HEK293 lysate.

Quality

Evaluated by Western Blotting on 10 mg of HEK293 cell lysate.

Target description

190 kDa Observed

Physical form

Format: Purified
Purified immunoglobulin in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Analysis Note

Control
HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Rictor on 10 µg of HEK293 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Wang et al.
Neoplasia (New York, N.Y.), 23(11), 1147-1165 (2021-10-28)
Epithelial-mesenchymal transition (EMT) contributes to tumor invasion, metastasis and drug resistance. AKT activation is key in a number of cellular processes. While many positive regulators for either EMT or AKT activation have been reported, few negative regulators are established. Through
Sharon K Kuss-Duerkop et al.
PLoS pathogens, 13(9), e1006635-e1006635 (2017-09-28)
Influenza A virus usurps host signaling factors to regulate its replication. One example is mTOR, a cellular regulator of protein synthesis, growth and motility. While the role of mTORC1 in viral infection has been studied, the mechanisms that induce mTORC1
Mahefatiana Andrianifahanana et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(11), 3733-3744 (2016-11-03)
TGF-β plays a central role in the pathogenesis of fibroproliferative disorders. Defining the exact underlying molecular basis is therefore critical for the development of viable therapeutic strategies. Here, we show that expression of the facilitative glucose transporter 1 (GLUT1) is
Mandy K Losiewicz et al.
Experimental eye research, 197, 108131-108131 (2020-07-06)
The retina is one of the most metabolically active tissues, yet the processes that control retinal metabolism remains poorly understood. The mTOR complex (mTORC) that drives protein and lipid biogenesis and autophagy has been studied extensively in regards to retinal
Sanjit K Dhar et al.
Oncogene, 37(48), 6225-6242 (2018-07-25)
Autophagy is a highly regulated evolutionarily conserved metabolic process induced by stress and energy deprivation. Here, we show that DNA polymerase gamma (Polγ) deficiency activates a selective prosurvival autophagic response via mitochondria-mediated reactive oxygen species (ROS) signaling and the mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service