Skip to Content
Merck
All Photos(1)

Key Documents

U-004

Supelco

U-47700-D3 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19D3Cl2N2O
Molecular Weight:
332.28
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

drug control

estupefaciente (Spain)

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

−20°C

General description

This stable-labeled Snap-N-Shoot® internal standard is offered as DEA-exempt solution and suitable for quantitation of U-47700 levels in urine or blood by LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications. U-47700 is a highly potent opioid agonist, not structurally related to fentanyl, that has been confirmed in opioid overdoses and deaths. Novel synthetic opioids (NSOs) created to circumvent drug control laws such as U-47700 have no recognized therapeutic use, are clandestinely manufactured and sold on conventional or dark web.

Legal Information

Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service