Skip to Content
Merck
All Photos(1)

Key Documents

S-065

Supelco

AB-FUBINACA solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21FN4O2
CAS Number:
Molecular Weight:
368.40
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)C[n]2nc(c3c2cccc3)C(=O)N[C@@H](C(C)C)C(=O)N

InChI

1S/C20H21FN4O2/c1-12(2)17(19(22)26)23-20(27)18-15-5-3-4-6-16(15)25(24-18)11-13-7-9-14(21)10-8-13/h3-10,12,17H,11H2,1-2H3,(H2,22,26)(H,23,27)/t17-/m0/s1

InChI key

AKOOIMKXADOPDA-KRWDZBQOSA-N

General description

AB-FUBINACA is a potent cannabinoid receptor agonist sold as a synthetic cannabinoid in Spice herbal blends. This certified reference solution is suited for use in synthetic cannabinoid testing methods by LC/MS o GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service