Skip to Content
Merck
All Photos(1)

Key Documents

H-921

Supelco

α-Hydroxymidazolam-d4 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D4H9ClFN3O
Molecular Weight:
345.79
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c([2H])c(c(F)c1[2H])C2=NCc3cnc(CO)n3-c4ccc(Cl)cc24

InChI

1S/C18H13ClFN3O/c19-11-5-6-16-14(7-11)18(13-3-1-2-4-15(13)20)22-9-12-8-21-17(10-24)23(12)16/h1-8,24H,9-10H2/i1D,2D,3D,4D

InChI key

QHSMEGADRFZVNE-RHQRLBAQSA-N

General description

This internal standard is suitable for midazolam testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring, or forensic analysis applications.

It is an internal standard of α-hydroxymidazolam, a major metabolite of the benzodiazepine midazolam, formed upon its metabolism by cytochrome P450 3A (CYP3A) isoenzymes.

Application

The Certified Spiking Solution® is suitable for quantitative testing of α-Hydroxymidazolam by HPLC, GC/MS, or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and Cannabis potency or impurity profiling testing applications.

The Certified Reference Material (CRM) in solution can also be used as follows:

  • Liquid chromatography-tandem mass spectrometry (LC–MS/MS) method based determination of 12 psychotropic drugs and their metabolites in guinea pig hair samples
  • Multiresidue analysis of 18 benzodiazepines in human blood and hair samples using LC–MS/MS
  • Detection and estimation of 21 benzodiazepines and three z-drugs from human hair samples after their two-step extraction by LC–MS/MS
  • Simultaneous analysis of 54 benzodiazepine and three z-drug residues from human urine and blood samples by ultra high-performance liquid chromatography- triple quadrupole tandem mass spectrometry (UHPLC-QqQ-MS/MS) method
  • Analysis of 26 compounds, comprising 18 benzodiazepines, seven benzodiazepine metabolites, and one z-drug from dried blood spot (DBS) cards using LC-MS/MS

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heesang Lee et al.
Forensic science international, 255, 137-145 (2015-08-11)
Dried Blood Spot (DBS) has been used a blood extraction method for inherited metabolic disorder screening since 1960s. With introduction of LC-MS/MS, not only DBS could be used to analysis drugs in small blood volume, but in various fields, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service