Skip to Content
Merck
All Photos(1)

Key Documents

E-016

Supelco

Ethyl-β-D-glucuronide

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O7
CAS Number:
Molecular Weight:
222.19
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](C(O)=O)O[C@@H](OCC)[C@@H]1O

InChI

1S/C8H14O7/c1-2-14-8-5(11)3(9)4(10)6(15-8)7(12)13/h3-6,8-11H,2H2,1H3,(H,12,13)/t3-,4-,5+,6-,8+/m0/s1

InChI key

IWJBVMJWSPZNJH-UQGZVRACSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material is widely used in forensic analysis and clinical toxicology applications to determine blood alcohol content.

Ethyl-β-D-glucuronide (EtG) is a water-soluble and stable metabolite of ethanol, used as a routine biomarker in alcohol abuse monitoring. A very small fraction of ethanol undergoes degradation to EtG, and it can be detected in body fluids as well as hair.

Application

The certified reference solution (CRM) in solution can also be used as follows:

  • Multi-residue analysis of different alcohol metabolites in human blood plasma and urine samples by ultra-high performance liquid chromatography (UHPLC) combined with tandem mass spectrometry (MS/MS)
  • Optimization of a supercritical fluid chromatography-tandem mass spectrometric (SFC-MS/MS) method for the separation and quantification of highly polar drugs and their metabolites using an ion-exchange stationary phase from human urine samples
  • Evaluation of a paired ion electrospray ionization mass spectrometric (PIESI-MS) approach coupled with high performance liquid chromatographic (HPLC) procedure for the detection and measurement of drug metabolites in urine samples
  • Assess the impact of sample preparation of hair samples on the quantitative analysis of ethyl-β-D-glucuronide by ultra-high performance liquid chromatography-electrospray ionization-tandem mass spectrometry
  • Analysis of ethyl-β-D-glucuronide as a urinary biomarker in human urine samples by a benchtop gas chromatograph coupled with a mass spectrometer (GC-MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive detection of anionic metabolites of drugs by positive ion mode HPLC-PIESI-MS
Guo H, et al.
International Journal of Mass Spectrometry, 389, 14-25 (2015)
A simplified approach for determination of urinary ethyl glucuronide by gas chromatography-mass spectrometry
Ghosh S, et al.
Journal of analytical science and technology, 12, 1-9 (2021)
Melanie Muskovich et al.
Traffic injury prevention, 13(6), 537-543 (2012-11-10)
The restriction of zero tolerance when driving (ZTD) imposed on drivers in Switzerland means that they may drink no alcohol at all before driving and basically consume only moderate amounts of alcohol at any other time. The Institute of Traffic
J Wesley Boyd et al.
Journal of addiction medicine, 6(4), 243-246 (2012-10-17)
Many physicians are referred to state physician health programs (PHPs) for evaluation, monitoring, and treatment of mental health and substance use disorders. Most PHPs are "diversion" or "safe haven" programs, meaning that physicians who suffer from alcohol or drug problems
A versatile UHPLC-MSMS method for simultaneous quantification of various alcohol intake related compounds in human urine and blood
Monosik R and Dragsted LO
Analytical Methods : Advancing Methods and Applications, 8, 6865-6871 (2016)

Articles

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service