Skip to Content
Merck
All Photos(1)

Key Documents

900112P

Avanti

PGF1α-d9

Avanti Research - A Croda Brand, powder

Synonym(s):

Prostaglandin F1α-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27D9O5
CAS Number:
Molecular Weight:
365.55
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

package of 1 × 100 μg
package of 1 × 50 μg
package of 1 × 500 μg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@@H](O)CC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])[C@H](O)C1

General description

Prostaglandin F1α-d9 (PGF1α-d9) is a deuterated derivative of PGF1α.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (900112-50UG)
2 mL Amber Serum Vial with Stopper and Crimp Cap (900112-100UG)
2 mL Amber Serum Vial with Stopper and Crimp Cap (900112-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service