Skip to Content
Merck
All Photos(1)

Key Documents

880129P

Avanti

DSPE-PEG(2000) Biotin

Avanti Research - A Croda Brand 880129P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (880129P-10mg)
pkg of 1 × 100 mg (880129P-100mg)
pkg of 1 × 25 mg (880129P-25mg)
pkg of 1 × 50 mg (880129P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880129P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CCCC[C@](SC[C@@]1(NC(N2)=O)[H])([C@]12[H])[H])=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=

General description

DSPE-PEG(2000) biotin is functionalized polyethylene glycol (PEG) conjugated lipid.

Application

DSPE-PEG(2000) biotin has been:
  • used as a component of liposomes, to prepare lipid microbubbles
  • used in the control experiments, to test the influence of 2,4-dinitrophenyl (DNP)-PEG2000-PE conformation on binding affinity
  • used for capsule surface modification
  • added to the labelling reaction to facilitate immobilization of the samples on the streptavidin-coated microfluidic sample chambers

Biochem/physiol Actions

Biotin is a member of vitamin B family of vitamins. It maintains the health of nails and hair. Biotin is known to promote insulin function and controls blood sugar levels.

Packaging

5 mL Amber Glass Screw Cap Vial (880129P-100mg)
5 mL Amber Glass Screw Cap Vial (880129P-10mg)
5 mL Amber Glass Screw Cap Vial (880129P-25mg)
5 mL Amber Glass Screw Cap Vial (880129P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound assisted siRNA delivery using PEG-siPlex loaded microbubbles
Vandenbroucke RE, et al.
J. Controlled Release, 126(3), 265-273 (2008)
The Definitive Guide to Natural Pregnancy Health - Why Your Prenatal Vitamin May Not Be Enough, 8-8 (2007)
Wen-Qun Li et al.
International journal of nanomedicine, 14, 8345-8360 (2019-11-07)
The protective role of puerarin (PUE) against myocardial infarction is closely related to its regulation on mitochondria. However, free PUE can hardly reach the mitochondria of ischemic cardiomyocytes due to the lack of mitochondrial targeting of PUE. Here PUE was
Associating HIV-1 envelope glycoprotein structures with states on the virus observed by smFRET
Lu M, et al.
Nature, 568(7752), 415-415 (2019)
Natasha D Durham et al.
Viruses, 12(1) (2020-01-19)
The Ebola virus (EBOV) envelope glycoprotein (GP) mediates the fusion of the virion membrane with the membrane of susceptible target cells during infection. While proteolytic cleavage of GP by endosomal cathepsins and binding of the cellular receptor Niemann-Pick C1 protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service