Skip to Content
Merck
All Photos(1)

Key Documents

870262P

Avanti

18:1 Dodecanyl PE

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanyl) (sodium salt), powder

Synonym(s):

Sodium (2R)-2,3-bis{[(9Z)-octadec-9-enoyl]oxy}propyl 2-[(11-carboxyundecanoyl)amino]ethyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H97NO11PNa
CAS Number:
Molecular Weight:
978.30
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870262P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870262P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCCCCCCCC(O)=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C53H98NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-31-35-39-43-52(58)62-47-49(65-53(59)44-40-36-32-26-24-22-20-18-16-14-12-10-8-6-4-2)48-64-66(60,61)63-46-45-54-50(55)41-37-33-29-27-28-30-34-38-42-51(56)57;/h17-20,49H,3-16,21-48H2,1-2H3,(H,54,55)(H,56,57)(H,60,61);/q;+1/p-1/b19-17-,20-18-;/t49-;/m1./s1

InChI key

AHHCDNFLXOEQMC-VXILFBGRSA-M

General description

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl) (18:1 Dodecanyl PE) or N-dod-PE, is a fusogenic lipid, belonging to the class of head group modified functionalized lipids.

Application

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-dodecanoyl (18:1 Dodecanyl PE) may be used in preparation of fusogenic and unilamellar liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (870262P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rico C Gunawan et al.
Biomaterials, 31(5), 900-907 (2009-11-03)
Targeted drug delivery to inflamed or injured vascular endothelial cells (ECs) and smooth muscle cells (SMCs) may provide a precise and effective therapeutic treatment for cardiovascular diseases. Upregulation of cytokine-regulated cell surface receptors, intercellular cell adhesion molecule-1 (ICAM) and endothelial-leukocyte
T Shangguan et al.
Gene therapy, 7(9), 769-783 (2000-05-24)
A unique method for formulation of plasmid DNA with phospholipids has been devised for the purpose of producing vehicles that can mediate gene delivery and transfection of living cells. The polycation, spermine, was used to condense plasmid DNA within a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service