Skip to Content
Merck
All Photos(1)

Key Documents

860634P

Avanti

C16 Dihydroceramide (d18:0/16:0)

Avanti Research - A Croda Brand 860634P, powder

Synonym(s):

N-palmitoyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H69NO3
CAS Number:
Molecular Weight:
539.92
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860634P-10mg)
pkg of 1 × 5 mg (860634P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860634P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C34H69NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(37)32(31-36)35-34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h32-33,36-37H,3-31H2,1-2H3,(H,35,38)/t32-,33+/m0/s1

InChI key

GCGTXOVNNFGTPQ-JHOUSYSJSA-N

Application

C16 Dihydroceramide (d18:0/16:0) is suitable for use to study the potential relevance of the changes observed in the sphingolipid composition of autophagosomes and autolysosomes in tetrahydrocannabinol (THC) treated cells or to destabilize biological membranes.

Biochem/physiol Actions

Dihydroceramide acts as a precursor for ceramide synthesis. It has an ability to induce autophagy in the perinuclear region. Dihydroceramide affects autophagic flux and enhances the formation lipid droplets. Accumulation of C16-dihydroceramides might lead to liver injury.

Packaging

5 mL Amber Glass Screw Cap Vial (860634P-10mg)
5 mL Amber Glass Screw Cap Vial (860634P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dihydroceramide accumulation mediates cytotoxic autophagy of cancer cells via autolysosome destabilization
Hernandez-Tiedra S, et al.
Autophagy, 12(11), 2213-2229 (2016)
Md Abdullah Al Sazzad et al.
Langmuir : the ACS journal of surfaces and colloids, 32(23), 5973-5980 (2016-05-25)
To better understand the interactions of saturated ceramides with unsaturated glycerophospholipids in bilayer membranes, we measured how palmitoyl ceramide (PCer) and dihydroceramide (dihydro-PCer, lacking the trans 4 double bond of the sphingoid base of ceramide) can interact with phosphatidylcholines (PCs)
Dihydroceramide is a key metabolite that regulates autophagy and promotes fibrosis in hepatic steatosis model
Lee Ah young, et al.
Biochemical and biophysical research communications, 494(3-4), 460-469 (2017)
Sonia Hernández-Tiedra et al.
Autophagy, 12(11), 2213-2229 (2016-11-02)
Autophagy is considered primarily a cell survival process, although it can also lead to cell death. However, the factors that dictate the shift between these 2 opposite outcomes remain largely unknown. In this work, we used Δ9-tetrahydrocannabinol (THC, the main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service