858125P
Avanti
18:1 Lyso PG
1-oleoyl-2-hydroxy-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder
Synonym(s):
1-oleoy-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(18:1(9Z)/0:0); 110714
About This Item
Recommended Products
Assay
>99% (LPG; may contain up to 10% of the 2-LPG isomer, TLC)
form
powder
packaging
pkg of 1 × 1 g (858125P-1g)
pkg of 5 × 100 mg (858125P-500mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 858125P
shipped in
dry ice
storage temp.
−20°C
SMILES string
[H][C@@](COP([O-])(OCC(O)CO)=O)(O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]
InChI
1S/C24H47O9P.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-24(28)31-19-23(27)21-33-34(29,30)32-20-22(26)18-25;/h9-10,22-23,25-27H,2-8,11-21H2,1H3,(H,29,30);/q;+1/p-1/b10-9-;/t22?,23-;/m1./s1
InChI key
STLDIXSFXILIFT-AXFOVFEGSA-M
General description
Application
- for the preparation of liposomes
- to determine the effects of phosphoglycerol(PG) and cardiolipin (CL) in F0F1 ATPase c subunit (F0-c) transformation
- as an acyl acceptor to determine the substrate specificity of recombinant G0/G1 switch gene 2 (G0S2) for acyl acceptors
Biochem/physiol Actions
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service