Skip to Content
Merck
All Photos(2)

Key Documents

850105P

Avanti

20:4 Lyso PI

1-arachidonoyl-2-hydroxy-sn-glycero-3-phosphoinositol (ammonium salt), powder

Synonym(s):

1-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol); PI(20:4(5Z,8Z,11Z,14Z)/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H52NO12P
CAS Number:
Molecular Weight:
637.70
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (LPI; may contain up to 10% of the 2-LPI isomer, TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850105P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850105P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850105P

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(O)COC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O.[NH4+]

InChI

1S/C29H49O12P.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(31)39-20-22(30)21-40-42(37,38)41-29-27(35)25(33)24(32)26(34)28(29)36;/h6-7,9-10,12-13,15-16,22,24-30,32-36H,2-5,8,11,14,17-21H2,1H3,(H,37,38);1H3/b7-6-,10-9-,13-12-,16-15-;/t22-,24-,25-

InChI key

QUUPQGFXDAUQSE-BIEDJLKKSA-N

General description

20:4 Lyso PI or 1-arachidonoyl-2-hydroxy-sn-glycero-3-phosphoinositol (ammonium salt) is an ammonium salt of phosphoinositol, with substitution of arachidonic acid at the sn-1 position of the glycerol backbone.
Increased circulating levels of L-alpha-lysophosphatidylinositol (LPI) are associated with cancer and LPI is a potent ligand for the G-protein-coupled receptor GPR55.

Application

20:4 Lyso PI or has been used as a lipid standard for optimization of the liquid chromatography mass spectrometry (LC-MS) method conditions.[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850105P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850105P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate
Jung Yong Eum et al.
Journal of chromatography. A, 460849-460849 (2020-01-14)
Aging refers to the intracellular accumulation of reactive oxygen species that damages proteins, DNA, and lipids. As alterations in lipid metabolism may trigger metabolic disorders and the onset of metabolic diseases, changes in lipid profiles can be closely related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service