Skip to Content
Merck
All Photos(2)

Key Documents

840885P

Avanti

18:2 PA

1,2-dilinoleoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1,2-di-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphate (sodium salt); PA(18:2(9Z,12Z)/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68O8PNa
CAS Number:
Molecular Weight:
718.92
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840885P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840885P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O.[Na+]

InChI

1S/C39H69O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h11-14,17-20,37H,3-10,15-16,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b13-11-,14-12-,19-17-,20-18-;/t37-;/m1./s1

InChI key

UMYNPOZOTCNQKB-DKSSBCNHSA-M

General description

Phosphatidic acid (PA) is a precursor of lysophosphatidic acid (LPA) and is present in leafy vegetables.

Application

18:2 PA or 1,2-dilinoleoyl-sn-glycero-3-phosphate (sodium salt) might be used in saposin-derived lipid nanoparticle (SapNP) formation and in vacuolar mimic lipid (VML) proteoliposomes used for content-mixing assays.

Packaging

5 mL Clear Glass Sealed Ampule (840885P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Flayhan et al.
Structure (London, England : 1993), 26(2), 345-355 (2018-02-08)
Saposin-derived lipid nanoparticles (SapNPs) are a new alternative tool for membrane protein reconstitution. Here we demonstrate the potential and advantages of SapNPs. We show that SapA has the lowest lipid specificity for SapNP formation. These nanoparticles are modular and offer
Hongki Song et al.
eLife, 9 (2020-01-22)
Yeast vacuole fusion requires R-SNARE, Q-SNAREs, and HOPS. A HOPS SM-family subunit binds the R- and Qa-SNAREs. We now report that HOPS binds each of the four SNAREs. HOPS catalyzes fusion when the Q-SNAREs are not pre-assembled, ushering them into
Mohamad Navab et al.
Journal of lipid research, 54(12), 3403-3418 (2013-10-03)
We recently reported that levels of unsaturated lysophosphatidic acid (LPA) in the small intestine significantly correlated with the extent of aortic atherosclerosis in LDL receptor-null (LDLR⁻/⁻) mice fed a Western diet (WD). Here we demonstrate that WD increases unsaturated (but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service