Skip to Content
Merck
All Photos(1)

Key Documents

810528C

Avanti

16:0 DNP Cap PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[6-[(2,4-dinitrophenyl)amino]hexanoyl] (ammonium salt), chloroform

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-[6-[(2,4-dinitrophenyl)amino]caproyl] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H90N5O13P
CAS Number:
Molecular Weight:
988.24
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (810528C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810528C

concentration

10 mg/mL (810528C-25mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoethanolamine is a precursor to phosphatidylcholine and phosphatidylethanolamine. 2,4-dinitrophenol (DNP) is a synthetic lipid-soluble compound which transports ions across the bioenergetic membranes.

Application

16:0 DNP Cap PE may be used in Rho-PE-doped ink solution for the cell recruiting experiments. It may also be used to study its patterning conditions in order to pattern multiple species.

Biochem/physiol Actions

Phosphoethanolamine participates in phospholipid metabolism. It has the ability to repress tumor growth both in vitro and in vivo.

Packaging

5 mL Clear Glass Sealed Ampule (810528C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumonisin, Folate, and Neural Tube Defects
Comprehensive Toxicology, 12, 189-208 (2010)
Membrane Transport
Stillwell, William
An Introduction to Biological Membranes, 423-451 (2016)
Konstantinos Mitsakakis et al.
The Analyst, 137(13), 3076-3082 (2012-05-26)
The present work demonstrates for the first time patterning of a ready-to-use biosensor with several different biomolecules using Dip-Pen Nanolithography (DPN) for the development of a procedure towards more rapid and efficient multi-sample detection. The biosensor platform used is based
D W Ellison et al.
Brain research, 417(2), 389-392 (1987-08-11)
Measurements of both phosphoethanolamine (PEA) and ethanolamine (EA) were made in postmortem brain samples from patients with Alzheimer's disease (AD) and Huntington's disease (HD) using high-performance liquid chromatography with electrochemical detection. In AD levels of PEA were significantly reduced by
Jürgen Strasser et al.
ACS nano (2019-12-31)
Activation of membrane receptors through clustering is a common mechanism found in various biological systems. Spatial proximity of receptors may be transduced across the membrane to initiate signaling pathways or alternatively be recognized by peripheral proteins or immune cells to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service